Assay Details
| Gene Symbol: Jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033398.2 | NP_203971.2 | 5 - 6 | 1335 | 66 | |
| GenBank mRNA | AF304118.1 | - | 5 - 6 | 1268 | 66 | |
| BC006067.1 | - | 5 - 6 | 1144 | 66 | ||
| BC011503.1 | - | 5 - 6 | 1739 | 66 | ||
| BC056629.1 | - | 5 - 6 | 1335 | 5033487 | 66 | |
| BC117737.1 | - | 5 - 6 | 1070 | 66 | ||
| BP769727.1 | - | 1 - 2 | 66 | |||
| EF527404.1 | - | 6 - 7 | 1424 | 66 | ||
| EF527405.1 | - | 6 - 7 | 1172 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
kidney development
sprouting angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
cell surface receptor signaling pathway
multicellular organism development
heart development
RNA splicing
chromatin modification
peptidyl-lysine hydroxylation to 5-hydroxy-L-lysine
cell differentiation
lung development
T cell differentiation in thymus
macrophage activation
apoptotic cell clearance
recognition of apoptotic cell
regulation of mRNA splicing, via spliceosome
erythrocyte development
oxidation-reduction process
retina development in camera-type eye
histone H3-R2 demethylation
histone H4-R3 demethylation
sulfonate dioxygenase activity
RNA binding
single-stranded RNA binding
receptor activity
iron ion binding
protein binding
oxidoreductase activity
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
histone demethylase activity (H3-R2 specific)
histone demethylase activity (H4-R3 specific)
hypophosphite dioxygenase activity
identical protein binding
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity
peptidyl-lysine 5-dioxygenase activity
blood vessel development
kidney development
sprouting angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
cell surface receptor signaling pathway
multicellular organism development
heart development
RNA splicing
chromatin modification
peptidyl-lysine hydroxylation to 5-hydroxy-L-lysine
cell differentiation
lung development
T cell differentiation in thymus
macrophage activation
apoptotic cell clearance
recognition of apoptotic cell
regulation of mRNA splicing, via spliceosome
erythrocyte development
oxidation-reduction process
retina development in camera-type eye
histone H3-R2 demethylation
histone H4-R3 demethylation
sulfonate dioxygenase activity
RNA binding
single-stranded RNA binding
receptor activity
iron ion binding
protein binding
oxidoreductase activity
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
histone demethylase activity (H3-R2 specific)
histone demethylase activity (H4-R3 specific)
hypophosphite dioxygenase activity
identical protein binding
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity
peptidyl-lysine 5-dioxygenase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161569 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161573 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161571 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161570 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114543 | EF527405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114543 | BC056629.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114543 | BC117737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114543 | CX210791.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114543 | EF527404.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499524 | M84742.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499524 | NM_007723.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499524 | U19717.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AF203969.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK004317.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK010566.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK045238.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK149953.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK157205.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK158036.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK170842.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK171838.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | AK215308.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | BC006748.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | BC023297.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909403 | NM_018813.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10243 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158441 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114543 | EF527405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114543 | BC056629.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114543 | BC117737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114543 | CX210791.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114543 | EF527404.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P428721 | AK122317.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P428721 | BC051491.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332952 | BC056629.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332952 | BP769727.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332952 | AF304118.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332952 | BC006067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332952 | NM_033398.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P162466 | AK122317.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28161536 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158441 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07506523 | - | - |
| Whole Mouse Genome G4122A | A_52_P428721 | AK122317.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P428721 | BC051491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332952 | BC056629.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332952 | BP769727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332952 | AF304118.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332952 | BC006067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332952 | NM_033398.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162466 | AK122317.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132073 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114543 | EF527405.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114543 | BC056629.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114543 | BC117737.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114543 | CX210791.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114543 | EF527404.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158441 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09565391 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09565395 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161571 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161573 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161569 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161570 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28161566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28161546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09494008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09319772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09494011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09319766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28161552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393394 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454109_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420056_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420057_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393387 | - | - |
| Mouse Genome 430 2.0 Array | 1420057_at | - | - |
| Mouse Genome 430 2.0 Array | 1441602_at | - | - |
| Mouse Genome 430 2.0 Array | 1420056_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1420057_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454109_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420056_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1441602_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454109_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107817_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107817_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100039152.mm.1 | NM_033398.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039152.mm.1 | NM_033398.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039152.mm.1 | TC1100004127.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039151.mm.1 | NM_033398.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039151.mm.1 | NM_033398.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039151.mm.1 | TC1100004127.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039150.mm.1 | NM_033398.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039150.mm.1 | NM_033398.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039150.mm.1 | TC1100004127.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 95486_PM_at | - | - |
| Murine Genome U74v2 Set | 95486_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454109_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420057_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454109_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420056_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420056_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420057_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4060100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242377 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242377 | - | - |
| mouse-6 v1.1 (Illumina) | scl39292.4_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39292.4_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1242377 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242377 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39292.4_16-S | - | - |

Jmjd6
Genomic Map