Assay Details
| Gene Symbol: Necab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021546.3 | NP_067521.2 | 12 - 13 | 1040 | 93 | |
| GenBank mRNA | AK013520.1 | - | 11 - 12 | 967 | 93 | |
| AK044296.1 | - | 12 - 13 | 1032 | 93 | ||
| AK158225.1 | - | 12 - 13 | 1042 | 93 | ||
| BC126874.1 | - | 10 - 11 | 854 | 40044848 | 93 | |
| BC126875.1 | - | 10 - 11 | 854 | 40044849 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761103 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761104 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761107 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761106 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761153 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761151 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083168 | AK044296.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083168 | AK158225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083168 | NM_021546.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083168 | AB039948.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137625 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133296 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083168 | AK044296.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083168 | AK158225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083168 | NM_021546.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083168 | AB039948.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269103 | AK044296.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269103 | AK158225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269103 | NM_021546.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269103 | AB039948.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269103 | AK013520.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04747274 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133296 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04747256 | - | - |
| Whole Mouse Genome G4122A | A_51_P269103 | AK044296.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269103 | AK158225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269103 | NM_021546.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269103 | AB039948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269103 | AK013520.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114625 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133296 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761153 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761103 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761107 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761106 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21761104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21761051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01372091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02256242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01704351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02066006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02092993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01775226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01372088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02256244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02256245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02066010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01704339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01775232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01704349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02092992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02256246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01252519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02066007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02256232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21761159 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488784 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488773 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488774 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488779 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488775 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488772 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431946_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447907_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488771 | - | - |
| Mouse Genome 430 2.0 Array | 1431946_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1431946_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1447907_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447907_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56846_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56846_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041896.mm.1 | NM_021546.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041896.mm.1 | NM_021546.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041896.mm.1 | TC0200004941.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041899.mm.1 | NM_021546.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041899.mm.1 | NM_021546.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041899.mm.1 | TC0200004941.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041897.mm.1 | NM_021546.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041897.mm.1 | NM_021546.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041897.mm.1 | TC0200004941.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041898.mm.1 | NM_021546.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041898.mm.1 | NM_021546.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041898.mm.1 | TC0200004941.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1447907_x_at | - | - |
| Mouse Expression Set 430 | 1447907_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431946_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431946_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447907_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447907_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5700341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700341 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775695 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775695 | - | - |
| mouse-6 v1.1 (Illumina) | scl18474.14.1_84-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18474.14.1_84-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700341 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2775695 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775695 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18474.14.1_84-S | - | - |

Necab3
Genomic Map