Assay Details
| Gene Symbol: Necab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021546.3 | NP_067521.2 | 2 - 3 | 144 | 78 | |
| GenBank mRNA | AK013520.1 | - | 2 - 3 | 143 | 78 | |
| AK044296.1 | - | 2 - 3 | 136 | 78 | ||
| AK158225.1 | - | 2 - 3 | 146 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Necab3
Genomic Map