| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200042265.mm.1 |
NM_010579.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042265.mm.1 |
NM_010579.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042265.mm.1 |
TC0200004973.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042260.mm.1 |
NM_010579.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042260.mm.1 |
NM_010579.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042260.mm.1 |
TC0200004973.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042264.mm.1 |
NM_010579.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042264.mm.1 |
NM_010579.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042264.mm.1 |
TC0200004973.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042268.mm.1 |
NM_010579.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042268.mm.1 |
NM_010579.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042268.mm.1 |
TC0200004973.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042269.mm.1 |
NM_010579.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042269.mm.1 |
NM_010579.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042269.mm.1 |
TC0200004973.mm.1 |
MTA TC |