Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010923.3 | NP_035053.1 | 2 - 3 | 267 | 63 | |
| GenBank mRNA | AK003004.1 | - | 2 - 3 | 235 | 63 | |
| AK160626.1 | - | 2 - 3 | 235 | 63 | ||
| BC036984.1 | - | 2 - 3 | 225 | 4981517 | 63 | |
| X83570.1 | - | 2 - 3 | 222 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nnat
Genomic Map