Assay Details
| Gene Symbol: Fam222b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145430.2 | NP_663405.1 | 1 - 2 | 121 | 54 | |
| GenBank mRNA | AK047860.1 | - | 1 - 2 | 105 | 54 | |
| AK085728.2 | - | 1 - 2 | 128 | 54 | ||
| AK140562.1 | - | 1 - 2 | 105 | 54 | ||
| AK141119.1 | - | 1 - 2 | 83 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK085728.2, AK141119.1, NM_145430.2 transcripts.
Set Membership: |
|

Fam222b
Genomic Map