Assay Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291236.1 | NP_001278165.1 | 1 - 2 | 563 | 99 | |
| NM_001291237.1 | NP_001278166.1 | 2 - 3 | 271 | 99 | ||
| GenBank mRNA | AA671983.1 | - | 2 - 3 | 288 | 99 | |
| AK165432.1 | - | 510 | 99 | |||
| BC039182.1 | - | 1 - 2 | 533 | 3500908 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tlcd1
Genomic Map