Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144830.3 | NP_659079.1 | 6 - 7 | 750 | 55 | |
| GenBank mRNA | AK136954.1 | - | 2 - 3 | 1744 | 55 | |
| AK146019.1 | - | 6 - 7 | 751 | 55 | ||
| AK149650.1 | - | 5 - 6 | 994 | 55 | ||
| AK150285.1 | - | 6 - 7 | 720 | 55 | ||
| AK151842.1 | - | 6 - 7 | 726 | 55 | ||
| AK152741.1 | - | 6 - 7 | 720 | 55 | ||
| AK154217.1 | - | 6 - 7 | 751 | 55 | ||
| AK168437.1 | - | 6 - 7 | 694 | 56 | ||
| AK169081.1 | - | 6 - 7 | 693 | 55 | ||
| AK170900.1 | - | 6 - 7 | 721 | 55 | ||
| AK172282.1 | - | 6 - 7 | 699 | 55 | ||
| BC022145.1 | - | 5 - 6 | 798 | 5101508 | 55 | |
| BC033268.1 | - | 6 - 7 | 723 | 5037750 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem106a
Genomic Map