Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144830.3 | NP_659079.1 | 8 - 9 | 847 | 74 | |
| GenBank mRNA | AK136954.1 | - | 4 - 5 | 1841 | 74 | |
| AK146019.1 | - | 8 - 9 | 848 | 74 | ||
| AK149650.1 | - | 7 - 8 | 1091 | 74 | ||
| AK150285.1 | - | 8 - 9 | 817 | 74 | ||
| AK151842.1 | - | 8 - 9 | 823 | 74 | ||
| AK152741.1 | - | 8 - 9 | 817 | 74 | ||
| AK154217.1 | - | 8 - 9 | 848 | 74 | ||
| AK168437.1 | - | 8 - 9 | 791 | 74 | ||
| AK169081.1 | - | 8 - 9 | 790 | 74 | ||
| AK170900.1 | - | 8 - 9 | 818 | 74 | ||
| AK172282.1 | - | 8 - 9 | 796 | 74 | ||
| BC022145.1 | - | 7 - 8 | 895 | 5101508 | 74 | |
| BC033268.1 | - | 8 - 9 | 820 | 5037750 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037121 | BC022145.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037121 | BC033268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037121 | AK170900.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037121 | AK172282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037121 | AK169081.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120294 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100664 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037121 | BC022145.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037121 | BC033268.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037121 | AK170900.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037121 | AK172282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037121 | AK169081.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074739 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100664 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07467113 | - | - |
| Whole Mouse Genome G4122A | A_51_P444669 | BC022145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444669 | AK170900.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444669 | AK172282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444669 | AK169081.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444669 | AK152741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444666 | BC022145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444666 | BC033268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444666 | AK170900.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444666 | AK172282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444666 | AK169081.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111746 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100664 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09711937 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09711932 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09711927 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09711949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09009503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09711949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09109314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09176198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09109307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09710400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09148371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09047152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09662051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09605033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09047151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28074766 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10381448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381450 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435263_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425025_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10381445 | - | - |
| Mouse Genome 430 2.0 Array | 1435263_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425025_at | - | - |
| Mouse Genome 430 2.0 Array | 1435263_at | - | - |
| Mouse Genome 430 2.0 Array | 1425025_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217203_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217203_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100015644.mm.1 | NM_144830.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015644.mm.1 | NM_144830.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015644.mm.1 | TC1100001661.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100015642.mm.1 | NM_144830.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015642.mm.1 | NM_144830.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015642.mm.1 | TC1100001661.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100015643.mm.1 | NM_144830.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015643.mm.1 | NM_144830.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015643.mm.1 | TC1100001661.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100015645.mm.1 | NM_144830.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015645.mm.1 | NM_144830.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015645.mm.1 | TC1100001661.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425025_PM_at | - | - |
| Mouse Expression Set 430 | 1425025_at | - | - |
| Mouse Expression Set 430 | 1435263_at | - | - |
| Mouse Expression Set 430 | 1435263_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425025_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425025_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425025_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425025_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435263_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435263_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4010348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4010348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004010348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238346 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238346 | - | - |
| mouse-6 v1.1 (Illumina) | scl40877.8.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4010348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40877.8.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4010348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1238346 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238346 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40877.8.1_11-S | - | - |

Tmem106a
Genomic Map