Assay Details
| Gene Symbol: Ptges3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026865.2 | NP_081141.1 | 3 - 4 | 899 | 89 | |
| GenBank mRNA | AK004415.1 | - | 3 - 4 | 217 | 89 | |
| AK007198.1 | - | 3 - 4 | 898 | 89 | ||
| BC057970.1 | - | 3 - 4 | 205 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptges3l
Genomic Map