Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026428.2 | NP_080704.2 | 2 - 3 | 207 | 89 | |
| GenBank mRNA | AK004023.1 | - | 2 - 3 | 206 | 89 | |
| AK153521.1 | - | 2 - 3 | 188 | 89 | ||
| AK161732.1 | - | 163 | 89 | |||
| BC012247.1 | - | 2 - 3 | 196 | 4189635 | 89 | |
| D89656.1 | - | 2 - 3 | 155 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcxr
Genomic Map