Assay Details
| Gene Symbol: Rac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133223.4 | NP_573486.1 | 4 - 5 | 423 | 80 | |
| GenBank mRNA | AB040819.1 | - | 4 - 5 | 290 | 80 | |
| AK089930.1 | - | 4 - 5 | 424 | 80 | ||
| AK150203.1 | - | 4 - 5 | 428 | 81 | ||
| AY974601.1 | - | 3 - 4 | 262 | 80 | ||
| BC025842.1 | - | 3 - 4 | 295 | 80 | ||
| BC117028.1 | - | 4 - 5 | 421 | 40126347 | 80 | |
| BC119041.1 | - | 4 - 5 | 421 | 8733790 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rac3
Genomic Map