Assay Details
| Gene Symbol: Snhg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027887.1 | 1 - 2 | 114 | 86 | ||
| GenBank mRNA | AK007971.1 | - | 1 - 2 | 115 | 86 | |
| AK040893.1 | - | 1 - 2 | 106 | 86 | ||
| AK079109.1 | - | 1 - 2 | 115 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lipopolysaccharide binding protein | Lbp |
Bpifd2, Ly88 |
-28.1kb | Anti-sense | Chr. 2 - 158306493 - 158332852 on NCBI Build GRCm38 |
| Downstream | small nucleolar RNA host gene 11 | Snhg11 |
A930034L06Rik, AI854265, AL022637, AW319638, E130013N09Rik |
14.1kb | Anti-sense | Chr. 2 - 158375638 - 158386145 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Snhg17
Genomic Map