Assay Details
| Gene Symbol: Kcnh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037712.1 | NP_001032801.1 | 2 - 3 | 305 | 74 | |
| GenBank mRNA | BC109174.1 | - | 2 - 3 | 305 | 40048550 | 74 |
| BC109175.1 | - | 2 - 3 | 305 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh6
Genomic Map