Assay Details
| Gene Symbol: Kcnh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037712.1 | NP_001032801.1 | 9 - 10 | 2150 | 120 | |
| GenBank mRNA | AK038513.1 | - | 3 - 4 | 649 | 120 | |
| BC109174.1 | - | 9 - 10 | 2150 | 40048550 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh6
Genomic Map