Assay Details
| Gene Symbol: Cct2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007636.2 | NP_031662.2 | 2 - 3 | 185 | 65 | |
| GenBank mRNA | AB041570.1 | - | 2 - 3 | 157 | 65 | |
| AK012295.1 | - | 2 - 3 | 141 | 65 | ||
| AK075581.1 | - | 2 - 3 | 158 | 65 | ||
| AK088145.1 | - | 2 - 3 | 148 | 65 | ||
| AK145406.1 | - | 2 - 3 | 186 | 65 | ||
| AK166891.1 | - | 2 - 3 | 158 | 65 | ||
| AK181482.1 | - | 2 - 3 | 96 | 65 | ||
| BC007470.1 | - | 2 - 3 | 149 | 3601569 | 65 | |
| BC026918.1 | - | 2 - 3 | 151 | 4500896 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cct2
Genomic Map