Assay Details
| Gene Symbol: Large | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010687.1 | NP_034817.1 | 1 - 2 | 90 | 64 | |
| GenBank mRNA | AJ006278.1 | - | 1 - 2 | 90 | 64 | |
| AK133940.1 | - | 1 - 2 | 574 | 64 | ||
| BC079596.1 | - | 1 - 2 | 375 | 64 | ||
| BC100399.1 | - | 1 - 2 | 153 | 30725031 | 64 | |
| BC125487.1 | - | 1 - 2 | 56 | 40130002 | 64 | |
| BC125489.1 | - | 1 - 2 | 56 | 40130004 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ006278.1, BC100399.1, BC125487.1, BC125489.1, NM_010687.1 transcripts.
Set Membership: |
|

Large
Genomic Map