Assay Details
| Gene Symbol: Large | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010687.1 | NP_034817.1 | 9 - 10 | 1307 | 73 | |
| GenBank mRNA | AJ006278.1 | - | 9 - 10 | 1307 | 73 | |
| AK122328.1 | - | 10 - 11 | 2384 | 73 | ||
| AK133940.1 | - | 9 - 10 | 1792 | 73 | ||
| BC061506.1 | - | 8 - 9 | 1064 | 73 | ||
| BC079596.1 | - | 9 - 10 | 1590 | 73 | ||
| BC100399.1 | - | 9 - 10 | 1370 | 30725031 | 73 | |
| BC125487.1 | - | 9 - 10 | 1273 | 40130002 | 73 | |
| BC125489.1 | - | 9 - 10 | 1273 | 40130004 | 73 | |
| HM141820.1 | - | 7 - 8 | 942 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Large
Genomic Map