Assay Details
| Gene Symbol: Banp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110100.2 | NP_001103570.1 | 4 - 5 | 586 | 75 | |
| NM_016812.4 | NP_058092.2 | 4 - 5 | 586 | 75 | ||
| GenBank mRNA | AK203167.1 | - | 1 - 2 | 179 | 75 | |
| AK208082.1 | - | 1 - 2 | 162 | 75 | ||
| AK219011.1 | - | 1 - 2 | 162 | 75 | ||
| BC021650.1 | - | 4 - 5 | 504 | 3660187 | 75 | |
| BC022168.1 | - | 4 - 5 | 504 | 4189797 | 75 | |
| BC062641.1 | - | 4 - 5 | 531 | 5718330 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Banp
Genomic Map