Assay Details
| Gene Symbol: Sntb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009229.4 | NP_033255.1 | 3 - 4 | 996 | 70 | |
| GenBank mRNA | AK040423.1 | - | 3 - 4 | 997 | 70 | |
| AK080059.1 | - | 3 - 4 | 993 | 70 | ||
| AK131687.1 | - | 3 - 4 | 1021 | 70 | ||
| AK165009.1 | - | 3 - 4 | 998 | 70 | ||
| AK165900.1 | - | 3 - 4 | 973 | 70 | ||
| BC030352.1 | - | 4 - 5 | 70 | |||
| BC038704.1 | - | 4 - 5 | 70 | |||
| BC051571.1 | - | 2 - 3 | 70 | |||
| BC138850.1 | - | 3 - 4 | 995 | 8861872 | 70 | |
| BC145442.1 | - | 3 - 4 | 995 | 9054000 | 70 | |
| U00678.1 | - | 3 - 4 | 995 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sntb2
Genomic Map