Assay Details
| Gene Symbol: Def8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253783.1 | NP_001240712.1 | 1 - 2 | 98 | 59 | |
| GenBank mRNA | BC003306.1 | - | 1 - 2 | 49 | 3154712 | 59 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003306.1, NM_001253783.1 transcripts.
Set Membership: |
|

Def8
Genomic Map