Assay Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172150.4 | NP_742162.2 | 1 - 2 | 61 | 59 | |
| GenBank mRNA | AK088467.1 | - | 1 - 2 | 62 | 59 | |
| AK160745.1 | - | 1 - 2 | 66 | 59 | ||
| AK167194.1 | - | 1 - 2 | 77 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088467.1, AK160745.1, AK167194.1, NM_172150.4 transcripts.
Set Membership: |
|

Ift52
Genomic Map