Assay Details
| Gene Symbol: Adgrg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033468.3 | NP_001028640.2 | 3 - 4 | 222 | 95 | |
| NM_001145972.1 | NP_001139444.1 | 3 - 4 | 227 | 95 | ||
| GenBank mRNA | AK028878.1 | - | 3 - 4 | 223 | 95 | |
| BC144851.1 | - | 3 - 4 | 188 | 9053399 | 95 | |
| BC145803.1 | - | 3 - 4 | 188 | 40131106 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgrg5
Genomic Map