Assay Details
| Gene Symbol: Gm11454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082432.1 | - | 1 - 2 | 148 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gametocyte specific factor 1-like | Gtsf1l | 2410116G06Rik | -33.0kb | Anti-sense | Chr. 2 - 163087031 - 163089601 on NCBI Build GRCm38 |
| Downstream | TOX high mobility group box family member 2 | Tox2 |
AI851523, AV026525, BI987407, Gcx1, RxHMG1 |
58.4kb | Sense | Chr. 2 - 163202643 - 163323102 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11454
Genomic Map