Assay Details
| Gene Symbol: Kctd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301173.1 | NP_001288102.1 | 3 - 4 | 497 | 79 | |
| NM_177791.3 | NP_808459.1 | 3 - 4 | 497 | 79 | ||
| GenBank mRNA | AK076644.1 | - | 3 - 4 | 489 | 79 | |
| BC092530.1 | - | 3 - 4 | 497 | 6744085 | 79 | |
| BC125431.1 | - | 3 - 4 | 489 | 40129946 | 79 | |
| BC125433.1 | - | 3 - 4 | 489 | 40129948 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kctd19
Genomic Map