Assay Details
| Gene Symbol: Acta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272041.1 | NP_001258970.1 | 2 - 3 | 172 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001272041.1 transcripts.
Set Membership: |
|

Acta1
Genomic Map