Assay Details
| Gene Symbol: Wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016873.2 | NP_058569.2 | 4 - 5 | 845 | 62 | |
| GenBank mRNA | AF100778.1 | - | 4 - 5 | 792 | 62 | |
| AF126063.1 | - | 4 - 5 | 777 | 62 | ||
| BC032877.1 | - | 4 - 5 | 770 | 1067632 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837605 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86889 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135199 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134218 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21814597 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134218 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135199 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04770588 | - | - |
| Whole Mouse Genome G4122A | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P390804 | BC032877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P390804 | NM_016873.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P390804 | AF100778.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129861 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135199 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01242810 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01242812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01242813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21814611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01545553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02071362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01242594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02071365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01242598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01872113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01272053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02071363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01678578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01242812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02079786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01272052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02079792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02079787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01545554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01872114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01900968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02027033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21814727 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10478418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478419 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419015_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478415 | - | - |
| Mouse Genome 430 2.0 Array | 1419015_at | - | - |
| Mouse Genome 430 2.0 Array | 1419015_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22403_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22403_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200020381.mm.1 | NM_016873.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020381.mm.1 | NM_016873.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020381.mm.1 | TC0200002496.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200020380.mm.1 | NM_016873.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020380.mm.1 | NM_016873.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020380.mm.1 | TC0200002496.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419015_at | - | - |
| Mouse Expression Set 430 | 1419015_PM_at | - | - |
| Murine Genome U74v2 Set | 94704_at | - | - |
| Murine Genome U74v2 Set | 94704_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419015_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419015_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419015_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419015_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 624707 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5550411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241893 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241893 | - | - |
| mouse-6 v1.1 (Illumina) | scl19952.5.1_24-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550411 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19952.5.1_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550411 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2486906 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2486906 | - | - |

Wisp2
Genomic Map