Assay Details
| Gene Symbol: Tomm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291155.1 | NP_001278084.1 | 1 - 2 | 256 | 89 | |
| GenBank mRNA | AK017699.1 | - | 1 - 2 | 201 | 89 | |
| AK075873.1 | - | 1 - 2 | 253 | 89 | ||
| AK168329.1 | - | 1 - 2 | 190 | 89 | ||
| BB869739.1 | - | 1 - 2 | 257 | 89 | ||
| BC018278.1 | - | 1 - 2 | 186 | 4187356 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tomm34
Genomic Map