Assay Details
| Gene Symbol: Tmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145439.2 | NP_663414.3 | 10 - 11 | 1452 | 59 | |
| GenBank mRNA | AK077671.1 | - | 8 - 9 | 1197 | 59 | |
| AK090179.1 | - | 10 - 11 | 1409 | 59 | ||
| AY236497.1 | - | 10 - 11 | 1351 | 59 | ||
| AY263158.1 | - | 10 - 11 | 1377 | 59 | ||
| BC004840.1 | - | 10 - 11 | 1377 | 2648849 | 59 | |
| BC005510.1 | - | 3 - 4 | 422 | 59 | ||
| BC006956.1 | - | 8 - 9 | 1162 | 59 | ||
| BC013502.1 | - | 1 - 2 | 179 | 4223599 | 59 | |
| BC058195.1 | - | 10 - 11 | 1377 | 5356348 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmc6
Genomic Map