Assay Details
| Gene Symbol: Sulf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252578.1 | NP_001239507.1 | 8 - 9 | 1624 | 84 | |
| NM_001252579.1 | NP_001239508.1 | 8 - 9 | 1470 | 84 | ||
| NM_028072.5 | NP_082348.2 | 8 - 9 | 1466 | 84 | ||
| GenBank mRNA | AK028874.1 | - | 8 - 9 | 1381 | 84 | |
| AK034712.1 | - | 8 - 9 | 1451 | 84 | ||
| AK036685.1 | - | 8 - 9 | 1402 | 84 | ||
| AK049170.1 | - | 3 - 4 | 381 | 84 | ||
| AK081643.1 | - | 8 - 9 | 1424 | 84 | ||
| AK129316.1 | - | 8 - 9 | 1624 | 84 | ||
| AK133336.1 | - | 1440 | 84 | |||
| AK165183.1 | - | 8 - 9 | 1378 | 84 | ||
| AY101177.1 | - | 7 - 8 | 1285 | 84 | ||
| BC062900.1 | - | 8 - 9 | 1423 | 84 | ||
| BC141086.1 | - | 7 - 8 | 1219 | 9055698 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sulf2
Genomic Map