Assay Details
| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011201.3 | NP_035331.3 | 2 - 3 | 319 | 113 | |
| GenBank mRNA | AK149979.1 | - | 2 - 3 | 275 | 113 | |
| AK150322.1 | - | 2 - 3 | 275 | 113 | ||
| AK151371.1 | - | 2 - 3 | 232 | 113 | ||
| AK153088.1 | - | 2 - 3 | 258 | 113 | ||
| AK156281.1 | - | 2 - 3 | 280 | 113 | ||
| AK157431.1 | - | 2 - 3 | 320 | 113 | ||
| AK165567.1 | - | 2 - 3 | 284 | 113 | ||
| AK165949.1 | - | 3 - 4 | 399 | 113 | ||
| AK166020.1 | - | 2 - 3 | 275 | 113 | ||
| AK168772.1 | - | 2 - 3 | 274 | 113 | ||
| AK170620.1 | - | 2 - 3 | 275 | 113 | ||
| AK171380.1 | - | 2 - 3 | 275 | 113 | ||
| AK172206.1 | - | 3 - 4 | 392 | 113 | ||
| AK185938.1 | - | 1 - 2 | 26 | 113 | ||
| BC010191.1 | - | 2 - 3 | 237 | 3156854 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK172206.1 transcripts.
Set Membership: |
|

Ptpn1
Genomic Map