Assay Details
| Gene Symbol: Ano4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277188.1 | NP_001264117.1 | 5 - 6 | 949 | 115 | |
| NR_102339.1 | 4 - 5 | 844 | 115 | |||
| GenBank mRNA | AK039466.1 | - | 5 - 6 | 751 | 115 | |
| AK039728.1 | - | 5 - 6 | 759 | 115 | ||
| AK078364.1 | - | 5 - 6 | 713 | 115 | ||
| BC065114.1 | - | 4 - 5 | 844 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ano4
Genomic Map