Assay Details
| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081381.1 | NP_001074850.1 | 1 - 2 | 161 | 57 | |
| GenBank mRNA | AK075986.1 | - | 1 - 2 | 158 | 57 | |
| BC048732.1 | - | 1 - 2 | 136 | 6512688 | 57 | |
| CJ073544.1 | - | 1 - 2 | 161 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P311905 | AK011270.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P311905 | BE448281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P311905 | NM_001081381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P311905 | AK011270.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P311905 | BE448281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P311905 | NM_001081381.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303343 | AF162685.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303343 | AK141422.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303343 | AK161577.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303343 | BC053026.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303343 | NM_016695.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569688 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655131 | AK035610.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 310083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71491 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171691 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137770 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P311905 | AK011270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P311905 | BE448281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P311905 | NM_001081381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P311905 | AK011270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P311905 | BE448281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P311905 | NM_001081381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P311905 | AK011270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P311905 | BE448281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P311905 | NM_001081381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P311905 | AK011270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P311905 | BE448281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P311905 | NM_001081381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619880 | AK011270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619880 | AK075986.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619880 | BC048732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619880 | BM943556.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619880 | NM_001081381.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08050090 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08050094 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08050085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08050089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26822972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26823038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07809952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07694950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08134036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07600675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26823013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26823005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08157174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08034510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08134038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26823004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08174136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26823003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26823010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26823007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26822973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26823012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10589508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589509 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453296_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440826_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10589503 | - | - |
| Mouse Genome 430 2.0 Array | 1440826_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1440826_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453296_at | - | - |
| Mouse Genome 430 2.0 Array | 1453296_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72341_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72341_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900012130.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012130.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012130.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012126.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012126.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012126.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012132.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012132.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012132.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012129.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012129.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012129.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012127.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012127.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012127.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012123.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012123.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012123.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012131.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012131.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012131.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012124.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012124.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012124.mm.1 | TC0900001458.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012128.mm.1 | NM_001081381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012128.mm.1 | NM_001081381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012128.mm.1 | TC0900001458.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453296_PM_at | - | - |
| Mouse Expression Set 430 | 1453296_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440826_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453296_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453296_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440826_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 792024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251253 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251253 | - | - |

Elp6
Genomic Map