Assay Details
| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081381.1 | NP_001074850.1 | 2 - 3 | 233 | 67 | |
| GenBank mRNA | AK011270.1 | - | 3 - 4 | 389 | 67 | |
| AK075986.1 | - | 2 - 3 | 230 | 67 | ||
| BC048732.1 | - | 2 - 3 | 208 | 6512688 | 67 | |
| CJ073544.1 | - | 2 - 3 | 233 | 67 | ||
More Information
Set Membership: |
|

Elp6
Genomic Map