Assay Details
| Gene Symbol: Dus1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026824.4 | NP_081100.2 | 2 - 3 | 503 | 97 | |
| GenBank mRNA | AK037538.1 | - | 1 - 2 | 917 | 97 | |
| AK088243.1 | - | 2 - 3 | 504 | 97 | ||
| AK147967.1 | - | 2 - 3 | 702 | 97 | ||
| AK148307.1 | - | 2 - 3 | 702 | 97 | ||
| AK211792.1 | - | 1 - 2 | 207 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dus1l
Genomic Map