Assay Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082020.1 | - | 2 - 3 | 221 | 64 | |
| AK170159.1 | - | 2 - 3 | 216 | 64 | ||
| AK171246.1 | - | 2 - 3 | 222 | 64 | ||
More Information
Set Membership: |
|

Sectm1a
Genomic Map