Assay Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK170159.1 | - | 3 - 4 | 264 | 75 | |
| AK171246.1 | - | 3 - 4 | 270 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sectm1a| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK170159.1 | - | 3 - 4 | 264 | 75 | |
| AK171246.1 | - | 3 - 4 | 270 | 75 | ||
Set Membership: |
|