Assay Details
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081164.1 | NP_001074633.1 | 12 - 13 | 1291 | 89 | |
| NM_001256033.1 | NP_001242962.1 | 12 - 13 | 1294 | 89 | ||
| GenBank mRNA | AK122429.1 | - | 7 - 8 | 585 | 89 | |
| BC138373.1 | - | 12 - 13 | 1266 | 8861393 | 89 | |
| BC145259.1 | - | 12 - 13 | 1263 | 9053812 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otud4
Genomic Map