| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800006604.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006604.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006604.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006604.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006604.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006606.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006606.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006606.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006606.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006606.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006607.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006607.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006607.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006607.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006607.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006594.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006594.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006594.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006594.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006594.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006600.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006600.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006600.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006600.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006600.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006603.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006603.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006603.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006603.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006603.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006597.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006597.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006597.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006597.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006597.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006596.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006596.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006596.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006596.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006596.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006599.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006599.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006599.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006599.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006599.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006601.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006601.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006601.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006601.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006601.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006605.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006605.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006605.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006605.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006605.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006608.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006608.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006608.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006608.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006608.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006610.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006610.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006610.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006610.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006610.mm.1 |
TC0800000882.mm.1 |
MTA TC |