| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800006572.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006572.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006572.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006572.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006572.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006573.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006573.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006573.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006573.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006573.mm.1 |
TC0800000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006574.mm.1 |
NM_001081164.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006574.mm.1 |
NM_001081164.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006574.mm.1 |
NM_001256033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006574.mm.1 |
NM_001256033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006574.mm.1 |
TC0800000882.mm.1 |
MTA TC |