Assay Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK150439.1 | - | 7 - 8 | 1352 | 83 | |
| BC045204.1 | - | 2 - 3 | 472 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxk2| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK150439.1 | - | 7 - 8 | 1352 | 83 | |
| BC045204.1 | - | 2 - 3 | 472 | 83 | ||
Set Membership: |
|