Assay Details
| Gene Symbol: Cyth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112699.1 | NP_001106169.1 | 3 - 4 | 305 | 68 | |
| NM_001112700.1 | NP_001106170.1 | 3 - 4 | 245 | 68 | ||
| NM_011180.3 | NP_035310.2 | 3 - 4 | 305 | 68 | ||
| GenBank mRNA | AB013464.1 | - | 3 - 4 | 226 | 68 | |
| AF051337.1 | - | 2 - 3 | 173 | 68 | ||
| AK078502.1 | - | 216 | 68 | |||
| AK158216.1 | - | 3 - 4 | 245 | 68 | ||
| AK172296.1 | - | 3 - 4 | 250 | 68 | ||
| AK220302.1 | - | 3 - 4 | 227 | 68 | ||
| BC057974.1 | - | 3 - 4 | 189 | 5060913 | 68 | |
| CJ131185.1 | - | 3 - 4 | 305 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyth1
Genomic Map