Assay Details
| Gene Symbol: Cyth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112699.1 | NP_001106169.1 | 7 - 8 | 680 | 75 | |
| NM_001112700.1 | NP_001106170.1 | 7 - 8 | 620 | 75 | ||
| NM_011180.3 | NP_035310.2 | 7 - 8 | 680 | 75 | ||
| GenBank mRNA | AB013464.1 | - | 7 - 8 | 601 | 75 | |
| AB035538.1 | - | 7 - 8 | 649 | 75 | ||
| AF051337.1 | - | 6 - 7 | 548 | 75 | ||
| AK078502.1 | - | 590 | 75 | |||
| AK158216.1 | - | 7 - 8 | 620 | 75 | ||
| AK172296.1 | - | 7 - 8 | 625 | 75 | ||
| AK220302.1 | - | 7 - 8 | 602 | 75 | ||
| AY126516.1 | - | 7 - 8 | 649 | 75 | ||
| BC057974.1 | - | 7 - 8 | 564 | 5060913 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyth1
Genomic Map