| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_001113486.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_001113486.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_001113487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_001113487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_001113488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_001113488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_017380.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
NM_017380.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018643.mm.1 |
TC1100001956.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_001113486.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_001113486.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_001113487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_001113487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_001113488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_001113488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_017380.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
NM_017380.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018644.mm.1 |
TC1100001956.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_001113486.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_001113486.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_001113487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_001113487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_001113488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_001113488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_017380.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
NM_017380.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018645.mm.1 |
TC1100001956.mm.1 |
MTA TC |