Assay Details
| Gene Symbol: Atp5h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027862.1 | NP_082138.1 | 2 - 3 | 157 | 64 | |
| GenBank mRNA | AF354051.1 | - | 1 - 2 | 150 | 64 | |
| AK002376.1 | - | 2 - 3 | 157 | 64 | ||
| AK088617.1 | - | 1 - 2 | 152 | 64 | ||
| BC016547.1 | - | 2 - 3 | 155 | 2631309 | 64 | |
| BC081431.1 | - | 2 - 3 | 156 | 6408035 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5h
Genomic Map