Assay Details
| Gene Symbol: Gm11725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090182.1 | - | 2 - 3 | 299 | 173 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 235 | Tmem235 |
Cldn27, Gm12581, OTTMUSG00000007589 |
-6.2kb | Sense | Chr. 11 - 117859199 - 117865543 on NCBI Build GRCm38 |
| Downstream | suppressor of cytokine signaling 3 | Socs3 |
Cis3, Cish3, EF-10, Ef10, SSI-3, Ssi3 |
93.9kb | Anti-sense | Chr. 11 - 117966079 - 117970341 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm11725
Genomic Map