Assay Details
| Gene Symbol: 4930423D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015188.1 | - | 1 - 2 | 226 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 40811 |
Gm40811 | -28.7kb | Sense | Chr. 10 - 117145168 - 117149728 on NCBI Build GRCm38 | |
| Downstream | YEATS domain containing 4 | Yeats4 |
4930573H17Rik, B230215M10Rik, GAS41, NuBI-1 |
34.4kb | Anti-sense | Chr. 10 - 117215141 - 117224507 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

4930423D24Rik
Genomic Map