Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 2 - 3 | 1531 | 82 | |
| GenBank mRNA | AK039929.1 | - | 2 - 3 | 1448 | 82 | |
| AK051602.1 | - | 2 - 3 | 1489 | 82 | ||
| AK134054.1 | - | 2 - 3 | 1450 | 82 | ||
| AK147708.1 | - | 2 - 3 | 1502 | 82 | ||
| AK155087.1 | - | 2 - 3 | 1501 | 82 | ||
| AK220187.1 | - | 2 - 3 | 1303 | 82 | ||
| BC120900.1 | - | 2 - 3 | 1250 | 40085687 | 82 | |
| BC120901.1 | - | 2 - 3 | 1250 | 40085688 | 82 | |
| D85612.1 | - | 2 - 3 | 1247 | 82 | ||
| EU887626.1 | - | 2 - 3 | 1219 | 82 | ||
| EU887627.1 | - | 2 - 3 | 1219 | 82 | ||
| EU887628.1 | - | 2 - 3 | 1219 | 82 | ||
| EU887629.1 | - | 2 - 3 | 1243 | 82 | ||
| EU887630.1 | - | 2 - 3 | 1243 | 82 | ||
| EU887631.1 | - | 2 - 3 | 1243 | 82 | ||
| U28807.1 | - | 2 - 3 | 1210 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc3
Genomic Map