Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 3 - 4 | 1694 | 117 | |
| GenBank mRNA | AK039929.1 | - | 3 - 4 | 1611 | 117 | |
| AK051602.1 | - | 3 - 4 | 1652 | 117 | ||
| AK131919.1 | - | 1 - 2 | 106 | 117 | ||
| AK134054.1 | - | 3 - 4 | 1613 | 117 | ||
| AK147708.1 | - | 3 - 4 | 1665 | 117 | ||
| AK155087.1 | - | 3 - 4 | 1664 | 117 | ||
| AK220187.1 | - | 3 - 4 | 1466 | 117 | ||
| BC120900.1 | - | 3 - 4 | 1413 | 40085687 | 117 | |
| BC120901.1 | - | 3 - 4 | 1413 | 40085688 | 117 | |
| D85612.1 | - | 3 - 4 | 1410 | 117 | ||
| EU887626.1 | - | 3 - 4 | 1382 | 117 | ||
| EU887627.1 | - | 3 - 4 | 1382 | 117 | ||
| EU887628.1 | - | 3 - 4 | 1382 | 117 | ||
| EU887629.1 | - | 3 - 4 | 1406 | 117 | ||
| EU887630.1 | - | 3 - 4 | 1406 | 117 | ||
| EU887631.1 | - | 3 - 4 | 1406 | 117 | ||
| U28807.1 | - | 3 - 4 | 1373 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc3
Genomic Map