Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 4 - 5 | 1898 | 88 | |
| GenBank mRNA | AK131919.1 | - | 2 - 3 | 310 | 88 | |
| AK134054.1 | - | 4 - 5 | 1817 | 88 | ||
| AK147708.1 | - | 4 - 5 | 1868 | 88 | ||
| AK220187.1 | - | 4 - 5 | 1670 | 88 | ||
| BC021835.1 | - | 1 - 2 | 102 | 88 | ||
| BC120900.1 | - | 4 - 5 | 1617 | 40085687 | 88 | |
| BC120901.1 | - | 4 - 5 | 1617 | 40085688 | 88 | |
| D85612.1 | - | 4 - 5 | 1614 | 88 | ||
| EU887626.1 | - | 4 - 5 | 1586 | 88 | ||
| EU887627.1 | - | 4 - 5 | 1586 | 88 | ||
| EU887629.1 | - | 4 - 5 | 1610 | 88 | ||
| EU887630.1 | - | 4 - 5 | 1610 | 88 | ||
| U28807.1 | - | 4 - 5 | 1577 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc3
Genomic Map